siRNA Details
| Gene Symbol: LOC101929269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134308.1 | 2 | 592 | |||
| GenBank mRNA | BC038544.1 | - | 2 | 592 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929269| Gene Symbol: LOC101929269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134308.1 | 2 | 592 | |||
| GenBank mRNA | BC038544.1 | - | 2 | 592 | ||