siRNA Details
| Gene Symbol: KCTD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005268492.2 | XP_005268549.1 | 3 | 12871 | ||
| XM_005268493.2 | XP_005268550.1 | 3 | 12267 | |||
| XM_006714789.2 | XP_006714852.1 | 3 | 12931 | |||
| XM_011537671.2 | XP_011535973.1 | 5 | 12766 | |||
| GenBank mRNA | AK091723.1 | - | 1 | 160 | ||
| BC036491.1 | - | 1 | 1977 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ942276.1 | - | 1 | 598 | Chr. 5 - 144484081 - 144484925 [+] on Build GRCh38 | |

KCTD16
Genomic Map