siRNA Details
| Gene Symbol: PRICKLE2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045697.1 | 3 | 3886 | |||
| GenBank mRNA | AL832189.1 | - | 3 | 3886 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRICKLE2-AS1| Gene Symbol: PRICKLE2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045697.1 | 3 | 3886 | |||
| GenBank mRNA | AL832189.1 | - | 3 | 3886 | ||