siRNA Details
| Gene Symbol: ZNF316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278559.1 | NP_001265488.1 | 9 | 4663 | ||
| XM_006715630.3 | XP_006715693.1 | 8 | 4780 | |||
| GenBank mRNA | AL832182.1 | - | 1 | 882 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF316| Gene Symbol: ZNF316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278559.1 | NP_001265488.1 | 9 | 4663 | ||
| XM_006715630.3 | XP_006715693.1 | 8 | 4780 | |||
| GenBank mRNA | AL832182.1 | - | 1 | 882 | ||