siRNA Details
| Gene Symbol: LOC101927560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_119374.1 | 3 | 1318 | |||
| GenBank mRNA | AK097722.1 | - | 3 | 1318 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927560| Gene Symbol: LOC101927560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_119374.1 | 3 | 1318 | |||
| GenBank mRNA | AK097722.1 | - | 3 | 1318 | ||