siRNA Details
| Gene Symbol: CALML3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120496.1 | 4 | 2930 | |||
| NR_120497.1 | 3 | 2818 | ||||
| GenBank mRNA | AK128534.1 | - | 3 | 3344 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CALML3-AS1| Gene Symbol: CALML3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120496.1 | 4 | 2930 | |||
| NR_120497.1 | 3 | 2818 | ||||
| GenBank mRNA | AK128534.1 | - | 3 | 3344 | ||