siRNA Details
| Gene Symbol: LOC221272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK025312.1 | - | 1 | 1759 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG620043.1 | - | 1 | 487 | Chr. 6 - 111277679 - 111278507 [+] on Build GRCh38 | |

LOC221272
Genomic Map