siRNA Details
Gene Symbol: LOC158402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056358.1 | - | 1 | 1203 | ||
AK095652.1 | - | 2 | 1847 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC158402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056358.1 | - | 1 | 1203 | ||
AK095652.1 | - | 2 | 1847 |