siRNA Details
| Gene Symbol: OXR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098168.1 | - | 1 | 379 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB203805.1 | - | 1 | 379 | Chr. 8 - 106749730 - 106750292 [+] on Build GRCh38 | |

OXR1
Genomic Map