siRNA Details
| Gene Symbol: CRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK095449.1 | - | 1 | 2079 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG107970.1 | - | 1 | 499 | Chr. 17 - 1420779 - 1421308 [-] on Build GRCh38 | |

CRK
Genomic Map