siRNA Details
| Gene Symbol: TOR1AIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC024226.1 | - | 2 | 813 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TOR1AIP2| Gene Symbol: TOR1AIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC024226.1 | - | 2 | 813 | ||