siRNA Details
| Gene Symbol: DNM3OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038397.2 | 1 | 4453 | |||
| NR_103486.1 | 4 | 547 | ||||
| GenBank mRNA | AK021543.1 | - | 1 | 207 | ||
| DB261523.1 | - | 4 | 506 | |||
| FJ623959.1 | - | 1 | 4453 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AU117617.1 | - | 1 | 207 | Chr. 1 - 172139707 - 172140589 [-] on Build GRCh38 | |

DNM3OS
Genomic Map