siRNA Details
| Gene Symbol: LOC101927132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110650.1 | 6 | 2217 | |||
| GenBank mRNA | AK091370.1 | - | 6 | 2219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927132| Gene Symbol: LOC101927132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110650.1 | 6 | 2217 | |||
| GenBank mRNA | AK091370.1 | - | 6 | 2219 | ||