siRNA Details
| Gene Symbol: SNHG22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_117096.1 | 4 | 1153 | |||
| GenBank mRNA | AK123464.1 | - | 4 | 1153 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000589499.1 | - | 4 | 1153 | Chr. 18 - 49814023 - 49851059 [+] on Build GRCh38 | |

SNHG22
Genomic Map