siRNA Details
| Gene Symbol: SNHG22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_117096.1 | 4 | 1592 | |||
| GenBank mRNA | AK123464.1 | - | 4 | 1592 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNHG22| Gene Symbol: SNHG22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_117096.1 | 4 | 1592 | |||
| GenBank mRNA | AK123464.1 | - | 4 | 1592 | ||