siRNA Details
Gene Symbol: LOC101927829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110814.1 | 1 | 1002 | |||
GenBank mRNA | BC041892.1 | - | 1 | 1002 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000455626.1 | - | 1 | 1002 | Chr. 3 - 20342468 - 20350928 [+] on Build GRCh38 |
