siRNA Details
| Gene Symbol: LOC101927829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110814.1 | 1 | 1182 | |||
| GenBank mRNA | BC041892.1 | - | 1 | 1182 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000455626.1 | - | 1 | 1182 | Chr. 3 - 20342468 - 20350928 [+] on Build GRCh38 | |

LOC101927829
Genomic Map