siRNA Details
| Gene Symbol: CMTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC018586.1 | - | 1 | 1176 | ||
| BC028676.1 | - | 1 | 244 | |||
| Gene Symbol: CKLF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK301240.1 | - | 5 | 811 | ||

CMTM1
Genomic Map