siRNA Details
| Gene Symbol: ZNF483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011518300.2 | XP_011516602.1 | 6 | 9758 | ||
| XM_017014337.1 | XP_016869826.1 | 7 | 8731 | |||
| XM_017014338.1 | XP_016869827.1 | 7 | 8617 | |||
| GenBank mRNA | AK054857.1 | - | 1 | 400 | ||

ZNF483
Genomic Map