siRNA Details
| Gene Symbol: LINC00458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108063.1 | 3 | 603 | |||
| GenBank mRNA | BC017942.1 | - | 3 | 598 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CX167424.1 | - | 3 | 552 | Chr. 13 - 54126963 - 54132820 [-] on Build GRCh38 | |

LINC00458
Genomic Map