siRNA Details
Gene Symbol: LINC01539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040025.1 | 3 | 558 | |||
NR_040026.1 | 2 | 426 | ||||
GenBank mRNA | AK057336.1 | - | 2 | 426 | ||
AK058169.1 | - | 3 | 539 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040025.1 | 3 | 558 | |||
NR_040026.1 | 2 | 426 | ||||
GenBank mRNA | AK057336.1 | - | 2 | 426 | ||
AK058169.1 | - | 3 | 539 |