siRNA Details
| Gene Symbol: KIAA1328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136303.1 | 14 | 2060 | |||
| GenBank mRNA | AK130479.1 | - | 1 | 2093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA1328| Gene Symbol: KIAA1328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136303.1 | 14 | 2060 | |||
| GenBank mRNA | AK130479.1 | - | 1 | 2093 | ||