siRNA Details
| Gene Symbol: LOC101929504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125863.1 | 3 | 395 | |||
| GenBank mRNA | AF086258.1 | - | 3 | 395 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | W67625.1 | - | 396 | Chr. - - [-] on Build | ||

LOC101929504
Genomic Map