siRNA Details
| Gene Symbol: MAPRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143826.2 | NP_001137298.1 | 8 | 3894 | ||
| NM_001143827.2 | NP_001137299.1 | 8 | 4040 | |||
| NM_001256420.1 | NP_001243349.1 | 6 | 3854 | |||
| NM_014268.3 | NP_055083.1 | 7 | 3982 | |||
| NR_046177.1 | 7 | 3797 | ||||

MAPRE2
Genomic Map