siRNA Details
| Gene Symbol: MAPRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143826.2 | NP_001137298.1 | 8 | 3974 | ||
| NM_001143827.2 | NP_001137299.1 | 8 | 4120 | |||
| NM_001256420.1 | NP_001243349.1 | 6 | 3934 | |||
| NM_014268.3 | NP_055083.1 | 7 | 4062 | |||
| NR_046177.1 | 7 | 3877 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR597075.1 | - | 1 | 1527 | Chr. 18 - 35141741 - 35143462 [+] on Build GRCh38 | |

MAPRE2
Genomic Map