siRNA Details
| Gene Symbol: LINGO1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120361.1 | 1 | 113 | |||
| GenBank mRNA | BC038758.1 | - | 1 | 112 | ||
| BI463387.1 | - | 1 | 117 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINGO1-AS2| Gene Symbol: LINGO1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120361.1 | 1 | 113 | |||
| GenBank mRNA | BC038758.1 | - | 1 | 112 | ||
| BI463387.1 | - | 1 | 117 | |||