siRNA Details
| Gene Symbol: LINGO1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120361.1 | 2 | 204 | |||
| NR_120362.1 | 2 | 186 | ||||
| GenBank mRNA | BC038758.1 | - | 2 | 203 | ||
| BI463387.1 | - | 2 | 208 | |||
| HY007517.1 | - | 2 | 175 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINGO1-AS2| Gene Symbol: LINGO1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120361.1 | 2 | 204 | |||
| NR_120362.1 | 2 | 186 | ||||
| GenBank mRNA | BC038758.1 | - | 2 | 203 | ||
| BI463387.1 | - | 2 | 208 | |||
| HY007517.1 | - | 2 | 175 | |||