siRNA Details
| Gene Symbol: JAML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090409.1 | - | 6 | 2948 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK057581.1 | - | 1 | 485 | Chr. 11 - 118199715 - 118201810 [-] on Build GRCh38 | |

JAML
Genomic Map