siRNA Details
| Gene Symbol: LINC00368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120415.1 | 2 | 95 | |||
| GenBank mRNA | BC040271.1 | - | 2 | 95 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG720241.1 | - | 2 | 100 | Chr. 13 - 111095838 - 111103097 [+] on Build GRCh38 | |

LINC00368
Genomic Map