siRNA Details
| Gene Symbol: PARD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005252536.4 | XP_005252593.1 | 16 | 4780 | ||
| XM_011519584.2 | XP_011517886.1 | 16 | 4789 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PARD3| Gene Symbol: PARD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005252536.4 | XP_005252593.1 | 16 | 4780 | ||
| XM_011519584.2 | XP_011517886.1 | 16 | 4789 | |||