siRNA Details
| Gene Symbol: PARD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005252536.4 | XP_005252593.1 | 16 | 4378 | ||
| XM_011519584.2 | XP_011517886.1 | 16 | 4387 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR618944.1 | - | 1 | 437 | Chr. 10 - 34343875 - 34345047 [-] on Build GRCh38 | |

PARD3
Genomic Map