siRNA Details
Gene Symbol: ZKSCAN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278119.1 | NP_001265048.1 | 7 | 6103 | ||
NM_001278121.1 | NP_001265050.1 | 7 | 6050 | |||
NM_001278122.1 | NP_001265051.1 | 6 | 6023 | |||
NM_006298.3 | NP_006289.2 | 6 | 6009 | |||
XM_011514870.2 | XP_011513172.1 | 6 | 6098 | |||
XM_011514873.2 | XP_011513175.1 | 7 | 6209 | |||
XM_011514874.2 | XP_011513176.1 | 5 | 5506 | |||
XM_017011266.1 | XP_016866755.1 | 6 | 6013 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR606559.1 | - | 1 | 288 | Chr. 6 - 28157790 - 28159467 [+] on Build GRCh38 |
