siRNA Details
| Gene Symbol: ZKSCAN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278119.1 | NP_001265048.1 | 7 | 7071 | ||
| NM_001278121.1 | NP_001265050.1 | 7 | 7018 | |||
| NM_001278122.1 | NP_001265051.1 | 6 | 6991 | |||
| NM_006298.3 | NP_006289.2 | 6 | 6977 | |||
| XM_011514870.2 | XP_011513172.1 | 6 | 7066 | |||
| XM_011514873.2 | XP_011513175.1 | 7 | 7177 | |||
| XM_011514874.2 | XP_011513176.1 | 5 | 6474 | |||
| XM_017011266.1 | XP_016866755.1 | 6 | 6981 | |||

ZKSCAN8
Genomic Map