siRNA Details
| Gene Symbol: LOC100132686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133566.1 | 1 | 188 | |||
| GenBank mRNA | BC020894.1 | - | 1 | 188 | 4702473 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100132686| Gene Symbol: LOC100132686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133566.1 | 1 | 188 | |||
| GenBank mRNA | BC020894.1 | - | 1 | 188 | 4702473 | |