siRNA Details
| Gene Symbol: CYP19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC022896.1 | - | 3 | 324 | ||
| BC056258.1 | - | 2 | 233 | |||
| DA853862.1 | - | 2 | 228 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | S96437.1 | - | 1 | 663 | Chr. 15 - 51255415 - 51256271 [-] on Build GRCh38 | |

CYP19A1
Genomic Map