siRNA Details
| Gene Symbol: MOBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003090.2 | 6 | 3722 | |||
| NR_103504.1 | 7 | 4064 | ||||
| NR_103505.1 | 7 | 4102 | ||||
| GenBank mRNA | AK095118.1 | - | 1 | 795 | ||
| BC035141.1 | - | 1 | 488 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MOBP| Gene Symbol: MOBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003090.2 | 6 | 3722 | |||
| NR_103504.1 | 7 | 4064 | ||||
| NR_103505.1 | 7 | 4102 | ||||
| GenBank mRNA | AK095118.1 | - | 1 | 795 | ||
| BC035141.1 | - | 1 | 488 | |||