siRNA Details
Gene Symbol: LOC283387 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC032840.1 | - | 1 | 2192 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000499481.2 | - | 1 | 2185 | Chr. 12 - 57869835 - 57896482 [-] on Build GRCh38 |
