siRNA Details
| Gene Symbol: LOC283387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC032840.1 | - | 1 | 2138 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283387| Gene Symbol: LOC283387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC032840.1 | - | 1 | 2138 | ||