siRNA Details
| Gene Symbol: EXTL3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126027.1 | 2, 3 | 706 | |||
| GenBank mRNA | BC118569.1 | - | 2, 3 | 706 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EXTL3-AS1| Gene Symbol: EXTL3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126027.1 | 2, 3 | 706 | |||
| GenBank mRNA | BC118569.1 | - | 2, 3 | 706 | ||