siRNA Details
| Gene Symbol: HOTTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037843.3 | 2 | 680 | |||
| GenBank mRNA | AK093987.1 | - | 2 | 664 | ||
| GU724873.1 | - | 2 | 680 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOTTIP| Gene Symbol: HOTTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037843.3 | 2 | 680 | |||
| GenBank mRNA | AK093987.1 | - | 2 | 664 | ||
| GU724873.1 | - | 2 | 680 | |||