siRNA Details
Gene Symbol: HOTTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037843.3 | 2 | 206 | |||
GenBank mRNA | AK093987.1 | - | 2 | 190 | ||
GU724873.1 | - | 2 | 206 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HOTTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037843.3 | 2 | 206 | |||
GenBank mRNA | AK093987.1 | - | 2 | 190 | ||
GU724873.1 | - | 2 | 206 |