siRNA Details
Gene Symbol: IPO5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103741.1 | 5 | 4137 | |||
NR_103742.1 | 4 | 4062 | ||||
GenBank mRNA | BC065573.1 | - | 1 | 1513 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IPO5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103741.1 | 5 | 4137 | |||
NR_103742.1 | 4 | 4062 | ||||
GenBank mRNA | BC065573.1 | - | 1 | 1513 |