siRNA Details
| Gene Symbol: NR2F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005654.5 | NP_005645.1 | 3 | 3471 | ||
| XM_017009797.1 | XP_016865286.1 | 3 | 1464 | |||
| GenBank mRNA | AF087978.1 | - | 1 | 338 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR2F1| Gene Symbol: NR2F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005654.5 | NP_005645.1 | 3 | 3471 | ||
| XM_017009797.1 | XP_016865286.1 | 3 | 1464 | |||
| GenBank mRNA | AF087978.1 | - | 1 | 338 | ||