siRNA Details
| Gene Symbol: Gprin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048200.1 | - | 1 | 842 | ||
| AK082455.1 | - | 1 | 105 | |||
| BC066049.1 | - | 2 | 2491 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gprin2| Gene Symbol: Gprin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048200.1 | - | 1 | 842 | ||
| AK082455.1 | - | 1 | 105 | |||
| BC066049.1 | - | 2 | 2491 | |||