siRNA Details
Gene Symbol: 4930432J09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045953.1 | 3 | 920 | |||
GenBank mRNA | AK015289.1 | - | 3 | 874 | ||
AK052661.1 | - | 3 | 921 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930432J09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045953.1 | 3 | 920 | |||
GenBank mRNA | AK015289.1 | - | 3 | 874 | ||
AK052661.1 | - | 3 | 921 |