siRNA Details
| Gene Symbol: Dlg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046356.1 | - | 1 | 2208 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB318438.2 | - | 1 | 476 | Chr. 7 - 90478403 - 90479068 [+] on Build GRCm38 | |

Dlg2
Genomic Map