siRNA Details
| Gene Symbol: Gm15179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037976.1 | 4 | 570 | |||
| GenBank mRNA | AK039780.1 | - | 4 | 568 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB198141.2 | - | 4 | 576 | Chr. 1 - 75368212 - 75374992 [-] on Build GRCm38 | |

Gm15179
Genomic Map