siRNA Details
| Gene Symbol: Gm15179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037976.1 | 3, 4 | 458 | |||
| GenBank mRNA | AK039780.1 | - | 3, 4 | 456 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15179| Gene Symbol: Gm15179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037976.1 | 3, 4 | 458 | |||
| GenBank mRNA | AK039780.1 | - | 3, 4 | 456 | ||