siRNA Details
| Gene Symbol: Gm15179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037976.1 | 1 | 16 | |||
| GenBank mRNA | AK039780.1 | - | 1 | 14 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB634114.1 | - | 1 | 14 | Chr. 1 - 75368212 - 75375012 [-] on Build GRCm38 | |

Gm15179
Genomic Map