siRNA Details
| Gene Symbol: Gm5091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046164.1 | 5 | 605 | |||
| GenBank mRNA | AK082876.1 | - | 5 | 606 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728917.1 | - | 5 | 606 | Chr. 17 - 15232135 - 15240783 [-] on Build GRCm38 | |

Gm5091
Genomic Map